This product requires proof of identity and completion of the DEA Iodine End Use Declaration before it can be purchased. Please complete this form if you have not previously done so.
Product Image
Thermo Scientific™
Remel™ Gram Iodine
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R40056Each
R40077Each
R400575/Pk.
Catalog number R40056
Price (INR)
-
Quantity:
Each
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
Specifications
Certifications/ComplianceCE Marked
QuantityEach
Product TypeGram Iodine
Volume (Metric)250 mL
Unit SizeEach