Product Image
Thermo Scientific™

Sulfuric Acid 10% Decolorizer

Use Sulfuric Acid 5% Decolorizer as a reagent in acid-fast staining procedures to detect Cryptosporidium in clinical specimens.

Have Questions?
Catalog NumberQuantity
R40126Each
Catalog number R40126
Price (INR)
-
Quantity:
Each

Use Thermo Scientific™ Remel™ Sulfuric Acid 1% Decolorizer in the acid-fast staining procedure to detect Cryptosporidium in clinical specimens.

Cryptosporidium is an intestinal protozoan parasite that causes diarrhea in animals as well as humans. This strain causes infection in immunocompromised hosts, especially those with AIDS (Acquired Immune Deficiency Syndrome)1. A modified acid-fast stain procedure was described by Ma and Soave because of its simplicity and specificity to detect Cryptosporidium2,3.

Cryptosporidium oocysts are spherical 4-6 μm in diameter, and stain pink to red to purple with Kinyoun carbolfuchsin stain. Most will contain several dark staining granules. Yeast cells, which are most likely to be confused with Cryptosporidium, stain blue or green based on the counterstain used. Sulfuric Acid 10% decolorizes the primary stain.

Cryptosporidium is more difficult to detect in formed stool specimens. Stain time is increased, which allows the oocysts to stand out from the background material. The hot acid-fast stain maximizes detection and identification of Cryptosporidium in formed stools4.

  • Easy-to-use - Examine smear in the slides mounted with mounting medium and a coverslip by bright field microscopy (40 X objective) and oil immersion is used (100 X objective) to observe the internal morphology.
  • Easy-to-interpret - In case of positive test oocysts stain pink to red to purple, are 4-6 μm in diameter and round or oval in shape. One to four sporozoites maybe visible within the oocyst. In case of negative test the yeast cells and background material stain green.

This reagent contains 10% of sulfuric acid (CAS 7664-93-9) in demineralized water (CAS 7732-18-5).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Murray, P.R., E.J. Baron, J.H. Jorgensen, M.L. Landry, and M.A. Pfaller. 2007. Manual of Clinical Microbiology. 9th ASM Press, Washington, D.C.
  2. Ma, P. and R. Soave. 1983. J. Infect. Dis. 147:824-828.
  3. Garza, D. 1983. Laboratory Medicine. 14:283-286.
  4. Garcia, L.S. 2001. Diagnostic Medical Parasitology. 4th ASM Press, Washington, D.C.
Specifications
QuantityEach
Product TypeDecolorizer
Volume (Metric)250 mL/Bottle
Unit SizeEach