Remel™ QC-Slide™ Microsporidia Stain Control
Thermo Scientific™
Remel™ QC-Slide™ Microsporidia Stain Control
Remel QC-Slide Microsporidia Stain Control is a methanol-fixed quality control slide used for Microsporidium stain procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R4015610 Slides/Pk.
R4015845 Slide/Pack
Catalog number R40156
Price (INR)
-
Quantity:
10 Slides/Pk.
Remel QC-Slide Microsporidia Stain Control is a methanol-fixed quality control slide used for Microsporidium stain procedures.
Sample preserved in 10% Formalin or SAF Fixative.
Specifications
DescriptionMicrosporidia Stain Control
FormatGlass Slide
Packaging Type10 Slide/Pack
Quantity10 Slides/Pk.
Product TypeMicrosporidia Stain Control
Unit SizeEach