Culti-Loops™ Aeromonas hydrophila ATCC™ 7966™
Actual product may vary
Culti-Loops™ Aeromonas hydrophila ATCC™ 7966™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46010205 Loops/Pack
Catalog number R4601020
Price (INR)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®7966TM*
DescriptionAeromonas hydrophila
FormatCulti-Loops
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Quantity5 Loops/Pack
Strain DesignationsWDCM00036, [CDC 359-60, IAM 12460, NCIB 9240, NCMB 86, NCTC 8049, RH 250]
SpeciesAeromonas species
Unit SizeEach