Culti-Loops™ Haemophilus influenzae Type b ATCC™ 33533™
Actual product may vary
Culti-Loops™ Haemophilus influenzae Type b ATCC™ 33533™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46039005 Loops/Pack
Catalog number R4603900
Price (INR)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®33533TM*
DescriptionHaemophilus influenzae Type b ATCC™ 33533™; Beta-lactamase negative
FormatCulti-Loops
Product TypeCulti-Loop
Target Organism ClassFastidious
Test TypeQualitative QC
Quantity5 Loops/Pack
Strain DesignationsNA
Unit SizeEach