Culti-Loops™ Salmonella sp. serovar Abony NCTC 6017
Actual product may vary
Culti-Loops™ Salmonella sp. serovar Abony NCTC 6017
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46060075 Loops/Pack
Catalog number R4606007
Price (INR)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
DescriptionSalmonella sp. serovar Abony NCTC™ 6017
FormatCulti-Loops
Product TypeCulti-Loop
Target Organism ClassGram-negative Rods
Quantity5 Loops/Pack
Strain DesignationsCCM 4518 [NBRC 100797, NCTC 6017, CIP 80.39]
Unit SizeEach