Culti-Loops™ Staphylococcus xylosus ATCC™ 29971™
Actual product may vary
Culti-Loops™ Staphylococcus xylosus ATCC™ 29971™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46065025 Loops/Pack
Catalog number R4606502
Price (INR)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®29971TM*
DescriptionStaphylococcus xylosus ATCC™ 29971™
FormatCulti-Loops
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassCocci
Test TypeQualitative QC
Quantity5 Loops/Pack
Strain DesignationsKL 162 [DSM 20266, LMG 20217, CCUG 7324, CIP 81.66, HAMBI 2057, JCM 2418, NCTC 11043, NRRL B-14776]
Unit SizeEach