Culti-Loops™ Streptococcus salivarius serotype II ATCC™ 13419™
This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Streptococcus salivarius serotype II ATCC™ 13419™
Thermo Scientific™
Culti-Loops™ Streptococcus salivarius serotype II ATCC™ 13419™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46082225 Loops/Pack
Catalog number R4608222
Price (INR)
-
Request A Quote
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®13419TM*
DescriptionStreptococcus salivarius serotype II ATCC™ 13419™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsWDCM00013, C699 [S30D]
Product TypeCulti-Loop
Target Organism ClassCocci
Unit SizeEach