Culti-Loops™ Salmonella enterica subsp enterica serovar Choleraesuis ATCC™ 10708™
This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Salmonella enterica subsp enterica serovar Choleraesuis ATCC™ 10708™
Thermo Scientific™
Culti-Loops™ Salmonella enterica subsp enterica serovar Choleraesuis ATCC™ 10708™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46090015 Loops/Pack
Catalog number R4609001
Price (INR)
-
Request A Quote
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®10708TM*
DescriptionSalmonella enterica subsp. enterica serovar Choleraesuis ATCC™ 10708™
FormatCulti-Loops
Quantity5 Loops/Pack
Strain DesignationsETS 34 [CIP 104220]
Product TypeCulti-Loop
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Unit SizeEach