Culti-Loops™ Streptococcus pyogenes group A ATCC™ 21547™
Actual product may vary
Culti-Loops™ Streptococcus pyogenes group A ATCC™ 21547™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46092115 Loops/Pk.
Catalog number R4609211
Price (INR)
-
Quantity:
5 Loops/Pk.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®21547TM*
Certifications/ComplianceCulti-Loops are CE Marked.
DescriptionStreptococcus pyogenes group A
FormatCulti-Loops
Product TypeSingle Organism
Target Organism ClassCocci
Test TypeQualitative QC
Packaging Type5 Loops/Pack
Quantity5 Loops/Pk.
Strain DesignationsWDCM00025, S-43
Unit SizeEach