Culti-Loops™ Cellulosimicrobium cellulans ATCC™ BAA-1817™
Thermo Scientific™
Culti-Loops™ Cellulosimicrobium cellulans ATCC™ BAA-1817™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46094225 Loops/Pk.
Catalog number R4609422
Price (INR)
-
Quantity:
5 Loops/Pk.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®BAA-1817TM*
Certifications/ComplianceCulti-Loops are CE Marked.
DescriptionCellulosimicrobium cellulans
Packaging Type5 Loops/Pack
Quantity5 Loops/Pk.
Strain DesignationsDSM 46215
Product TypeSingle Organism
Target Organism ClassGram-positive Rods
Test TypeQualitative QC
Unit SizeEach