Culti-Loops™ Clostridioides difficle ATCC™ BAA 1873™
Thermo Scientific™
Culti-Loops™ Clostridioides difficle ATCC™ BAA 1873™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46094365 Loops/Pk.
Catalog number R4609436
Price (INR)
-
Quantity:
5 Loops/Pk.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®BAA-1873™*
Certifications/ComplianceCulti-Loops are CE Marked.
DescriptionClostridioides difficile
FormatCulti-Loops
Packaging Type5 Loops/Pack
Quantity5 Loops/Pk.
Strain DesignationsWDCM00054
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassAnaerobes
Test TypeQualitative QC
Unit SizeEach