This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Escherichia coli ATCC™ BAA-201™  
Thermo Scientific™
Culti-Loops™ Escherichia coli ATCC™ BAA-201™  
Perform quality control testing and performance evaluation of microbiological procedures with these ready-to-use bacteriological loops.
Have Questions?
Catalog NumberQuantity
R46094825 Loops/Pk.
Catalog number R4609482
Price (INR)
-
Quantity:
5 Loops/Pk.
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Escherichia coli ATCC™ BAA-201™ strain produces extended-spectrum β-lactamase (ESBL) TEM-3. This strain is recommended to be used as a positive control strain with Thermo Scientific™ Spectra™ ESBL Medium following the HAI surveillance programs.
The loop can be streaked directly onto  appropriate media. Alternatively, it may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
For In Vitro Diagnostic Use.  
Specifications
ATCC®BAA-201
DescriptionEscherichia coli ATCC BAA-201
FormatCulti-Loops
Packaging Type5 Loops/Pack
Quantity5 Loops/Pk.
Regulatory Status*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Strain DesignationsJ53 pCFF04
Product TypeCulti-Loops Quality Control Organism
SpeciesEscherichia species
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Unit SizeEach