This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ API™ 20E QC Set
Thermo Scientific™
Culti-Loops™ API™ 20E QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46510035 Loops/Organism
Catalog number R4651003
Price (INR)
-
Quantity:
5 Loops/Organism
Thermo Scientific™ Culti-Loops™ API 20E Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Enterobacter cloacae ATCC™ 13047™†
Escherichia coli ATCC™ 25922™†
Klebsiella pneumoniae subsp. pneumoniae ATCC™ 35657™†
Proteus mirabilis ATCC™ 35659™†
Stenotrophomonas maltophilia ATCC™ 51331™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®13047TM*
DescriptionAPI 20E QC Set/5
FormatCulti-Loops
Quantity5 Loops/Organism
Product TypeCulti-Loops QC Set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach