This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Kirby Bauer Susceptibility QC Set
Thermo Scientific™
Culti-Loops™ Kirby Bauer Susceptibility QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46530045 Loops/Organism
Catalog number R4653004
Price (INR)
-
Quantity:
5 Loops/Organism
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures
.
Includes:
Escherichia coli ATCC™ 25922™†
Escherichia coli ATCC™ 35218™†
Pseudomonas aeruginosa ATCC™ 27853™†
Staphylococcus aureus supsp. aureus ATCC™ 25923™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®25922TM*
DescriptionKirby Bauer Susceptibility QC Set/4
FormatCulti-Loops
Quantity5 Loops/Organism
Product TypeCulti-Loop
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach