Culti-Loops™ BBL™ Crystal™ ID Systems Anaerobe QC Set/4
Thermo Scientific™
Culti-Loops™ BBL™ Crystal™ ID Systems Anaerobe QC Set/4
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46550231 Kit
Catalog number R4655023
Price (INR)
-
Quantity:
1 Kit
This product has been discontinued. Consider individual strains found from the set: Mfr. Nos. R4603050 and R4605001. Thermo Scientific™ Culti-Loops™ BBL Crystal ID Systems Anaerobe Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Fusobacterium varium ATCC™ 27725™
Lactobacillus acidophilus ATCC™ 314™†
Parabacteroides distasonis ATCC™ 8503™†
Peptoniphilus asaccharolyticus ATCC™ 29743™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
DescriptionBBL Crystal ID Systems Anaerobe QC Set/4
Quantity1 Kit
Product TypeCulti-Loops QC Set
Unit Size-