This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Sensititre™ MIC Breakpoint QC Set
Thermo Scientific™
Culti-Loops™ Sensititre™ MIC Breakpoint QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46870205 x 5 Loops
Catalog number R4687020
Price (INR)
-
Quantity:
5 x 5 Loops
Thermo Scientific™ Culti-Loops™ Sensititre MIC Breakpoint Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Enterococcus faecalis ATCC™ 29212 ™†
Escherichia coli ATCC™ 35218 ™†
Escherichia coli ATCC™ 25922 ™†
Pseudomonas aeruginosa ATCC™ 27853 ™†
Staphylococcus aureus subsp. aureus ATCC™ 29213 ™†
Ordering Information:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®29212TM*
DescriptionSensititre™ MIC Breakpoint QC Set/5
FormatCulti-Loops
Packaging Type5 x 5 Loops/Pack
Quantity5 x 5 Loops
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach