Culti-Loops™ Sensititre™ AquaCulture MIC QC Set
Thermo Scientific™
Culti-Loops™ Sensititre™ AquaCulture MIC QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46870222 x 1 Loop
Catalog number R4687022
Price (INR)
-
Quantity:
2 x 1 Loop
Thermo Scientific™ Culti-Loops™ Sensititre Aquaculture MIC Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Aeromonas salmonicida subsp. salmonicida ATCC™ 33658 ™†
Escherichia coli ATCC™ 25922 ™†
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®33658TM*
DescriptionSensititre™ Aquaculture MIC QC Set/2
FormatCulti-Loops
Quantity2 x 1 Loop
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach