This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Culti-Loops™ Sensititre™ H. influenzae/S.pneumoniae MIC QC Set
Thermo Scientific™
Culti-Loops™ Sensititre™ H. influenzae/S.pneumoniae MIC QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog NumberQuantity
R46870253 x 5 Loops
Catalog number R4687025
Price (INR)
-
Quantity:
3 x 5 Loops
Thermo Scientific™ Culti-Loops™ Sensititre H. influenzae/S.pneumoniae MIC Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Haemophilus influenzae ATCC™ 49247™ †
Haemophilus influenzae ATCC™ 49766™†
Streptococcus pneumoniae ATCC™ 49619™ †
Ordering Information:
Individually sealed in a foil pouch and packaged five loops per package.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®49247TM*
DescriptionSensititre™ H. influenzae/S.pneumoniae MIC QC Set/3
FormatCulti-Loops
Quantity3 x 5 Loops
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQualitative QC
Unit SizeEach