Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Thermo Scientific™
Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4711300100 Tests/Kit
Catalog number R4711300
Price (INR)
-
Request A Quote
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Brevundimonas diminuta ATCC™ 19146™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial
  • 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®19146TM*
DescriptionBrevundimonas diminuta ATCC™ 19146™
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsFDA strain PCI 818 [CCUG 24715, DSM 1635, LMG 10743]
Product TypeQuality Control Organism
Target Organism ClassGram-positive Rods
Test TypeQuantitative QC
Unit SizeEach