This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Typhimurium ATCC™ 13311™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Typhimurium ATCC™ 13311™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Typhimurium ATCC™ 13311™
Thermo Scientific™
Quanti-Cult Plus™ Salmonella enterca subsp. enterca serovar Typhimurium ATCC™ 13311™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4716100100 Tests/Kit
Catalog number R4716100
Price (INR)
-
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 13311™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • Ready for rehydration
  • For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®13311TM*
DescriptionSalmonella enterica subsp. enterica serovar Typhimurium ATCC™ 13311™
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsNCTC74
Product TypeQuality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Unit SizeEach