Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™
This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™
Thermo Scientific™
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog NumberQuantity
R4717020100 Tests/Kit
Catalog number R4717020
Price (INR)
-
Request A Quote
Quantity:
100 Tests/Kit
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control proceduresbullets1 (end)
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®29737TM*
DescriptionStaphylococcus aureus subsp. aureus ATCC™ 29737™
FormatQuanti-Cult Plus
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsFDA
Product TypeQuality Control Organism
Target Organism ClassCocci
Test TypeQuantitative QC
Unit SizeEach