This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing QC Set
Thermo Scientific™
Quanti-Cult Plus™ Convenience Set, Growth Promotion Testing QC Set
Supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Have Questions?
Catalog NumberQuantity
R4721000100 Tests/Set
Catalog number R4721000
Price (INR)
-
Quantity:
100 Tests/Set
Thermo Scientific™ Quanti-Cult Plus Growth Promotion Testing Set is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
Includes:
10 preserved organism vials and 10 rehydration vials
Contents:
  • Aspergillus brasiliensis ATCC™ 16404™
  • Bacillus spizizenii ATCC™ 6633
  • Candida albicans ATCC™ 10231™
  • Clostridium sporogenes ATCC™ 11437™
  • Pseudomonas paraeruginosa ATCC™ 9027™
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®16404™
DescriptionGrowth Promotion Testing QC Set/5
FormatQuanti-Cult Plus
Packaging Type100 Test/Set
Quantity100 Tests/Set
Product TypeQC set
Target Organism ClassOrganism Sets
Test TypeQuantitative QC
Unit SizeEach