This product requires registration before it can be purchased. Please complete the registration form if you have not previously registered.
Product Image
Thermo Scientific™
WKITS™ Water QC Kit 1003
Thermo Scientific™ WKIT Water QC Kit 1003 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
Have Questions?
Catalog NumberQuantity
R47410032/Pk.
Catalog number R4741003
Price (INR)
-
Quantity:
2/Pk.
The Thermo Scientific™ WKIT Water QC Kit 1003 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • WaterBugs™ deliver <50 CFUs in a single inoculum to mimic water samples
Ordering Alerts:
Reagents are sufficient for one test/reconstituted vial.
Includes:
Contents: 10 preserved organism vials (5 per organism); 10 rehydration vials. Escherichia coli strain C1, Pseudomonas aeruginosa ATCC™ 27853™†
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
DescriptionWater QC Kit 1003
Quantity2/Pk.
Product TypeQuality Control Kit
Unit SizeEach