Product Image
Thermo Scientific™

TCG-ProbeSwabQC™

Contains cells of T. vaginalis, C. albicans, and G. vaginalis for QC of probe systems and direct antigen detection kits.
Have Questions?
Catalog NumberQuantity
R4851035 Loops/Pack
Catalog number R485103
Price (INR)
-
Quantity:
5 Loops/Pack
Thermo Scientific™ TCG-ProbeSwabQC™ contains cells of T. vaginalis, C. albicans, and G. vaginalis for QC of probe systems and direct antigen detection kits.
Stabilized microorganism impregnated swab packaged in individual foil envelope.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
DescriptionTCG-ProbeSwabQC™
FormatProbeSwabQC
Quantity5 Loops/Pack
Product TypeBactidisk
Target Organism ClassOrganism Sets
Unit SizeEach