Product Image
Thermo Scientific™
Remel™ Ultraviolet Light
The Thermo Scientific™ Remel Ultraviolet Light is a Woods lamp with longwave UV and 365nm wavelength.
Have Questions?
Catalog NumberLight Source
R51084UV-C
Catalog number R51084
Price (INR)
-
Light Source:
UV-C
Thermo Scientific™ Remel Ultraviolet Light is a Woods lamp with longwave UV and 365nm wavelength.
120V
Baxter
Specifications
Light SourceUV-C
Voltage120 V
Wavelength365 nm
Unit SizeEach