Product Image
Thermo Scientific™

Remel™ Gonopak

Thermo Scientific™ Remel Gonopak is a Whirl-pak bag with wire tab fold-over closure.
Have Questions?
Catalog NumberFormat
R55048Whirl-pak
Catalog number R55048
Price (INR)
-
Format:
Whirl-pak

Thermo Scientific™ Remel Gonopak is a Whirl-pak bag with wire tab fold-over closure.

For use with Carbon Dioxide Tablets (Nasco)

Specifications
DescriptionGonopak
Quantity10/Pk.
FormatWhirl-pak
Unit SizeEach