Product Image
Thermo Scientific™
Remel™ Calgiswab™ Type 2 Swab Collection and Transport System, Wood-shaft
Thermo Scientific™ Remel Calgiswab Swab Collection and Transport System is designed for nasopharyngeal specimen collection.
Have Questions?
Catalog NumberQuantity
R601501050/Pk.
Catalog number R6015010
Price (INR)
-
Quantity:
50/Pk.
Thermo Scientific™ Remel Calgiswab Swab Collection and Transport System is designed for nasopharyngeal specimen collection.
Manufacturer: Puritan
Specifications
DescriptionWood-shaft; 1/Envelope; 50/Pk.
For Use With (Application)Healthcare
Packaging1/Envelope
Quantity50/Pk.
Shaft MaterialWood
FormatSwab
Product TypeRemel Calgiswab
Unit SizeEach