Product Image
Thermo Scientific™
Remel™ Chlamydia-Pak™ Specimen Collection Kit
Specimen collection and transport kit for C. trachomatis direct fluorescent antibody tests (DFA).
Have Questions?
Catalog NumberQuantity
R623001 Test/Kit
Catalog number R62300
Price (INR)
-
Quantity:
1 Test/Kit
Thermo Scientific™ Remel Chlamydia-Pak Specimen Collection Kit is a specimen collection and transport kit for C. trachomatis direct fluorescent antibody tests (DFA).
Manufacturer: Medical Packaging Corporation
Includes:
One large dacron-tipped plastic shaft swab; Small dacron-tipped stainless steel shaft swab; Nylon bristle cytology brush; Single well glass slide; Ampule methanol fixative.
Specifications
For Use With (Application)Direct Fluorescent Antibody Tests (DFA)
Quantity1 Test/Kit
Product TypeTest Kit
Unit SizeEach