Product Image
Thermo Scientific™
Remel™ Microloop
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeCalibrated
R6419551/1000Yes
R641970SmallNo
R641980LargeNo
R641990MediumNo
R6419051/100Yes
Catalog number R641955
Price (INR)
-
Size:
1/1000
Calibrated:
Yes
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Specifications
CalibratedYes
Quantity1 Ea.
MaterialNichrome
Size1/1000
Volume0.001 μL
Unit SizeEach