BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Thermo Scientific™
BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingSwab Configuration
R72300550/Pk.Single Applicator
R723000500/Pk.Single Applicator
R723010500/Pk.Dual Applicator
R72301550/Pk.Dual Applicator
Catalog number R723005
Price (INR)
-
Packaging:
50/Pk.
Swab Configuration:
Single Applicator
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Blue cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Closure ColorBlue
DescriptionAmies Clear
Packaging50/Pk.
Quantity1 Tube/Envelope
Shaft MaterialPlastic
FormatTube
Media TypeAmies Clear
Swab ConfigurationSingle Applicator
Unit SizeEach