BactiSwab™: Wire Shaft Liquid Stuart
Thermo Scientific™
BactiSwab™: Wire Shaft Liquid Stuart
Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
R723130100/Pk.
R72313550/Pk.
Catalog number R723130
Price (INR)
-
Packaging:
100/Pk.
Thermo Scientific™ Remel BactiSwab: Wire Shaft Liquid Stuart is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
Rayon-tipped, aluminum-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
  • Straight aluminum shaft
  • Green cap
Specifications
Closure ColorGreen
DescriptionLiquid Stuarts
For Use With (Application)Healthcare
Packaging100/Pk.
Shaft MaterialWire
FormatTube
Media TypeLiquid Stuart
Unit SizeEach