Ramsey™ Series 20 Belt Scale System
Thermo Scientific™
Ramsey™ Series 20 Belt Scale System
The Thermo Scientific™ Ramsey™ Series 20 belt scale system monitors feed with an accuracy of ±0.5%, even in the harshest applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
RAMSEYFLEXINT-20Ramsey™ Flex Digitizer
RAMSEYFLEXDIG-20Ramsey™ Flex Integrator
14354Series 20 Belt Scale System
Catalog number RAMSEYFLEXINT-20
Price (INR)
-
Description:
Ramsey™ Flex Digitizer
At Thermo Fisher Scientific, we provide our valued customers with the products, solutions, and support that enable you to make the world healthier, cleaner, and safer.
To ensure we can continue to offer innovation, quality, value, and acceptable lead times we continuously evaluate our portfolio.
Aftermarket support, including remote technical support, select field services, and spare parts (subject to availability), may be available. Contact our service team for details.
Specifications
DescriptionRamsey™ Flex Digitizer
Unit SizeEach