AquaSensor™ DataStick™ Wide-Range Turbidity Measurement System
Actual product may vary
Thermo Scientific™
AquaSensor™ DataStick™ Wide-Range Turbidity Measurement System
Monitor wastewater and surface water turbidity with the Thermo Scientific™ AquaSensor™ DataStick™ Wide-Range Turbidity Measurement System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
RT1SSStainless steel
RT1CPVCCPVC
Catalog number RT1SS
Price (INR)
-
Description:
Stainless steel
Monitor wastewater and surface water turbidity with the Thermo Scientific™ AquaSensors™ DataStick™ Wide-Range Turbidity Measurement System, which features a replaceable, pre-calibrated (for turbidity and temperature) sensor head.
The measurement system provides plug-and-play capability into any Thermo Scientific™ DataStick™ communications adapter for interactive measurement, calibration, configuration and diagnostics with industrial computers or with the Thermo Scientific™ AquaSensors™ AV38 local display and connects directly to a PLC.
Turbidity Measurement System Components:
  • Wide-Range Water Turbidity Sensor Head
  • Communications Adapter
Wash Head:
  • For use in applications where the sensor head can be quickly fouled with sediment
  • Head slips on the sensor body and wash nozzle aligns with sensor fingers
  • Actuation controlled by AV38 wash relay programmed with wash interval and duration
AV38 Local Display/Controller:
  • Two-line display and seven key navigation
  • Data reporting with up to two current outputs
  • Two Form C relays
  • Digital communications
Recommended for:
  • Drinking water
  • Raw water monitoring
  • Back water monitoring
  • Wastewater effluent
  • Process clarity monitoring
Specifications
Accuracy1% of reading
ApprovalsMeets CE requirements for heavy industrial use
Certifications/ComplianceCE, UL, ISO
DescriptionStainless steel
Resolution0.1 NTU
Step Response Time90% in <15 sec.
Range0 to 4000 NTU
TypeWide-Range Turbidity Measurement System, Stainless Steel
Unit SizeEach