Product Image
Gibco™

Rat (Sprague Dawley) Microsomes

Rat (Sprague Dawley) Microsomes
Have Questions?
Catalog NumberQuantity
RTMCPL0.5 mL
Catalog number RTMCPL
Price (INR)
3,832.00
온라인 행사
Ends: 30-Sep-2026
6,966.00
Save 3,134.00 (45%)
Each
Quantity:
0.5 mL
Price (INR)
3,832.00
온라인 행사
Ends: 30-Sep-2026
6,966.00
Save 3,134.00 (45%)
Each
Liver microsomes prepared from a pool of male Sprague Dawley rats.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 mg/mL
Donor SourcePooled
GenderMale
Quantity0.5 mL
Shipping ConditionDry Ice
Sub Cellular LocalizationMicrosomes
Target SpeciesRat
TissueLiver
FormFrozen
Product LineGibco
Product TypeSubcellular Fractions
Unit SizeEach
Contents & Storage
Store between -60°C and -85°C