Store Image and Database Management Software, enterprise edition
Store Image and Database Management Software, enterprise edition
Thermo Scientific™
Store Image and Database Management Software, enterprise edition
Thermo Scientific™ Store Image and Database Management Software, Enterprise Edition, is used for managing the images and data automatically transferredRead more
Have Questions?
Catalog number S0100015
Price (INR)
5,780,871.00
Each
Add to cart
Price (INR)
5,780,871.00
Each
Add to cart
Thermo Scientific™ Store Image and Database Management Software, Enterprise Edition, is used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product. It allows for rapid, highly scalable, secure, storage of images and data with the ability to query.
Features of the Store Image and Database Management Software, Enterprise Edition:
  • Automatic Transfer of Images and Data—Centralized data storage allowing data and image access from multiple locations
  • Accessible and Linked—Data is seamlessly linked to its associated image
  • Visualize—View images and data linked together and easily compare images
  • Export—Built in export formats for software such as Spotfire™, Excel™, De Novo™ Software, and GeneData Screener™ programs
The Store Image and Database Management Software maintains data and images on a SQL™ or Oracle™ database.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 each
For Use With (Application)Data/Image Management
For Use With (Equipment)HCA/HCS
LicenseEnterprise
Operating SystemCellInsight
Product TypeImage Management, Database Management
Unit SizeEach