Store Image and Database Management Software, standard edition
Thermo Scientific™
Store Image and Database Management Software, standard edition
Thermo Scientific™ Store Image and Database Management Software, Standard Edition, is used for managing the images and data automatically transferredRead more
Have Questions?
Catalog number S0100015D
Price (INR)
2,610,271.00
Each
Price (INR)
2,610,271.00
Each
Thermo Scientific™ Store Image and Database Management Software, Standard Edition, is used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product. It allows for rapid, highly scalable, secure, storage of images and data with the ability to query.
Features of the Store Image and Database Management Software, Standard Edition:
• Automatic Transfer of Images and Data—Centralized data storage allowing data and image access from multiple locations
• Accessible and Linked—Data is seamlessly linked to its associated image
• Visualize—View images and data linked together and easily compare images
• Export—Built in export formats for software such as Spotfire™, Excel™, De Novo™ Software, and GeneData Screener™ programs
The Store Image and Database Management Software maintains data and images on a SQL™ or Oracle™ database.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionStore Informatics Software Standard Edition
For Use With (Equipment)ArrayScan High Content Analysis (HCA) Readers, CellInsight CX5 High Content Screening (HCS) Platform
LicenseStandard
Quantity1 each
SoftwareImage Management, Database Management
Unit SizeEach