SiteClick™ Duet and Quartet Antibody Labeling Kits
SiteClick™ Duet and Quartet Antibody Labeling Kits
SiteClick™ Duet and Quartet Antibody Labeling Kits
SiteClick™ Duet and Quartet Antibody Labeling Kits
SiteClick™ Duet and Quartet Antibody Labeling Kits
Invitrogen™

SiteClick™ Duet and Quartet Antibody Labeling Kits

Introducing SiteClick Duet and Quartet kits. Experience the same robust, covalent, site-specific antibody conjugation you trust, now with a faster and easier workflow. Complete in less than 24 hours with fewer workflow steps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
S11001Quartet Antibody Modification Kit5 mg
S11002Quartet Antibody Modification Kit1 mg
S11003Quartet Antibody Modification Kit100 μg
S11004Duet Antibody Modification Kit5 mg
S11005Duet Antibody Modification Kit1 mg
S11006Duet Antibody Modification Kit100 μg
S11007Quartet Antibody Modification Kit3 x 100 μg
S11008Duet Antibody Modification Kit3 x 100 μg
Catalog number S11001
Price (INR)
293,733.00
Each
Product Type:
Quartet Antibody Modification Kit
Quantity:
5 mg
Price (INR)
293,733.00
Each

SiteClick Duet and Quartet antibody labeling kits are used to create label-ready, site-specific, azido-modified antibodies without lengthy genetic modifications. Our enzyme-driven SiteClick labeling precisely attaches an azido moiety to the heavy chains of IgG antibodies, preserving antigen-binding domains for optimal target binding. This site selectivity targets carbohydrate domains on the Fc region, applicable to all IgG antibodies regardless of isotype or host species.

The SiteClick Quartet Antibody Labeling Kit offers up to four payloads per antibody (DOS 4) in less than 24 hours, while the SiteClick Duet Antibody Labeling Kit enables the attachment of two payloads (DOS 2) in only 4 hours.

Features of the SiteClick Duet and Quartet antibody labeling kits include:

  • Modify any IgG antibody species or isotype with a complete kit
  • Follow a brief, straightforward protocol
  • Preserve antibody function with efficient, site-specific labeling
  • Achieve consistent results with reproducible labeling chemistry
  • Develop therapeutic antibodies, antibody-drug conjugates, antibody-oligo conjugates, bispecific antibodies, and immunodiagnostics

Once azido-modified, choose from a variety of SiteClick sDIBO or DBCO alkyne labels for covalent attachment via copper-free click chemistry under mild conditions. Enjoy the flexibility to select different fluorescent labels, including Alexa Fluor, pHrodo, and LysoLight dyes, or to conjugate other molecules such as biotin, oligos, toxins, or degraders.

SiteClick labeling is useful for developing therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostic tests, helping ensure high specificity, reproducible labeling, and preserved antibody performance.

Custom SiteClick Antibody Labeling Service

Unlock the full potential of your antibodies with our Custom SiteClick Antibody Labeling Service and customized labels. If conventional methods have failed or reduced your antibody's activity, our expert team can help. We offer SiteClick-compatible labels in bulk format, and custom labeling with a variety of detection molecules, including Alexa Fluor, LysoLight, and pHrodo dyes, as well as biotin and R-PE.

Interested in using SiteClick technology beyond your internal research? Let us help you use this in your product manufacturing and services. Please inquire about licensing options for your commercial applications.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Antibody Production and Labeling
For Use With (Equipment)Benchtop Centrifuge
FormatLabeling Kit
Green FeaturesImproved stability, less waste
Incubation Time16 to 20 hr.
Label TypeDegree of Labeling: up to 4
Labeling MethodClick Chemistry
Product LineSiteClick
Product TypeQuartet Antibody Modification Kit
Quantity5 mg
Reactive MoietyAzide
Shipping ConditionWet Ice
SpeciesAny
Sufficient For1 Labeling
Labeling TargetIgG, Antibodies
Unit SizeEach
Contents & Storage
2°C to 8°C

Citations & References (2)

Citations & References
Abstract
Oligonucleotide conjugated antibody strategies for cyclic immunostaining.
Authors:Gibbs SL
Journal:Scientific reports
PubMed ID:34903759
Synthesis and In Vivo Evaluation of a Site-specifically Labeled Radioimmunoconjugate for Dual-Modal (PET/NIRF) Imaging of MT1-MMP in Sarcomas.
Authors:Knight JC
Journal:Bioconjugate chemistry
PubMed ID:35867034