SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 680
Invitrogen™

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 680

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 680
Have Questions?
Catalog NumberQuantity
S300451 kit
Catalog number S30045
Price (INR)
-
Quantity:
1 kit

The Alexa Fluor™ 680 SAIVI™ Antibody Labeling Kit provides a convenient means to label antibodies with an optimal degree of labeling for in vivo imaging applications (DOL; ∼2) over a 6-fold antibody concentration range with no adjustments in reaction volume, dye concentration, or antibody concentration necessary. Using this procedure, optimally labeled antibodies are ready for applications that require azide-free reagents, such as live-cell imaging or direct injection into animals.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
ColorInfrared
Detection MethodFluorescence
Excitation/Emission679/702
Label TypeAlexa Fluor Dyes
Labeling Scale0.5 to 3 mg
Product LineAlexa Fluor, SAIVI
Product TypeLabeling Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Labeling TargetAntibodies
Label or DyeAlexa Fluor 680
Unit SizeEach
Contents & Storage
Store in refrigerator 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

What amount of conjugated antibody should I inject to image tumors?

A recommended starting dosage is 50 µg. You will need to determine the optimal dosage for your experimental model.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What size needle should I use for small animal in vivo imaging?

We recommend the use of a 28-32 gauge tuberculin or insulin syringe (0.3 or 1.0 mL volume) with a fixed (non-removable) needle.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the optimal and maximal volumes of reagent that can be injected into a mouse?

The volume of reagent that can be injected varies according to the route of administration. The following numbers are general guidelines for a 25 gram animal: Intravenous (IV)- 50-125 µl (recommended)- 200µl (maximum) ; Intraperitoneal (IP) 500µl (recommended) -2ml (maximum) ; Subcutaneous(SC) 100-250 µl (recommended)- 1ml (maximum).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What type of mice are the best for in vivo imaging?

Due to light scattering caused by fur, hairless mice such as athymic nude (nu/nu) mice are recommended for in vivo imaging. If this is not an option, the hair covering the area to be imaged should be removed using clippers or a chemical depilatory such as Nair depilatory.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (5)

Citations & References
Abstract
Affibody Molecules for In vivo Characterization of HER2-Positive Tumors by Near-Infrared Imaging.
Authors:Lee SB, Hassan M, Fisher R, Chertov O, Chernomordik V, Kramer-Marek G, Gandjbakhche A, Capala J,
Journal:Clin Cancer Res
PubMed ID:18559604
'PURPOSE: HER2 overexpression has been associated with a poor prognosis and resistance to therapy in breast cancer patients. We are developing molecular probes for in vivo quantitative imaging of HER2 receptors using near-infrared (NIR) optical imaging. The goal is to provide probes that will minimally interfere with the studied system, ... More
Noninvasive positron emission tomography and fluorescence imaging of CD133+ tumor stem cells.
Authors:Gaedicke S, Braun F, Prasad S, Machein M, Firat E, Hettich M, Gudihal R, Zhu X, Klingner K, Schüler J, Herold-Mende CC, Grosu AL, Behe M, Weber W, Mäcke H, Niedermann G,
Journal:
PubMed ID:24469819
'A technology that visualizes tumor stem cells with clinically relevant tracers could have a broad impact on cancer diagnosis and treatment. The AC133 epitope of CD133 currently is one of the best-characterized tumor stem cell markers for many intra- and extracranial tumor entities. Here we demonstrate the successful noninvasive detection ... More
Minimally invasive quantification of lymph flow in mice and rats by imaging depot clearance of near-infrared albumin.
Authors:Karlsen TV, McCormack E, Mujic M, Tenstad O, Wiig H,
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:22101523
'There is a lack of available methods to noninvasively quantify lymphatic function in small experimental animals, a necessity for studies on lymphatic system pathophysiology. We present a new method to quantify lymph flow in mice and rats, based on optically monitoring the depot clearance of near-infrared fluorescently labeled albumin and ... More
Multiplexed mAbs: a new strategy in preclinical time-domain imaging of acute myeloid leukemia.
Authors:McCormack E, Mujic M, Osdal T, Bruserud Ø, Gjertsen BT,
Journal:Blood
PubMed ID:23243270
Antibodies play a fundamental role in diagnostic immunophenotyping of leukemias and in cell-targeting therapy. However, this versatility is not reflected in imaging diagnostics. In the present study, we labeled anti–human mAbs monochromatically against selected human myeloid markers expressed on acute myeloid leukemia (AML) cells, all with the same near-infrared fluorochrome. ... More
Influenza virus aerosol exposure and analytical system for ferrets.
Authors:Gustin KM, Belser JA, Wadford DA, Pearce MB, Katz JM, Tumpey TM, Maines TR,
Journal:Proc Natl Acad Sci U S A
PubMed ID:21536880
Understanding the transmission ability of newly emerging influenza viruses is central to the development of public health preparedness and prevention strategies. Animals are used to model influenza virus infection and transmission, but the routinely used intranasal inoculation of a liquid virus suspension does not reflect natural infection. We report the ... More