Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts
Actual product may vary
Thermo Scientific™
Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts.
Have Questions?
Catalog Number
Product Type
S4S-DB
Buffer Drain Bottle
Catalog number S4S-DB
Price (INR)
-
Request A Quote
Product Type:
Buffer Drain Bottle
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
Specifications
For Use With (Equipment)Owl™ S4S Aluminum-Backed Sequencer System