IDEIA™ Chlamydia Specimen Wet Collection Kit
Thermo Scientific™
IDEIA™ Chlamydia Specimen Wet Collection Kit
Simplify the wet collection and transportation of the urethral and endocervical specimens to be tested with this collection kit.
Have Questions?
Catalog number S600730-2
Price (INR)
-
Simplify the wet collection and transportation of urethral and endocervical specimens to be tested with Thermo Scientific™ IDEIA™ Chlamydia Specimen Wet Collection Kit.
Chlamydiae are intracellular organisms that infect the columnar epithelial surfaces1 of the human urethra and endocervix. Specimens collected from these sites must contain as many columnar epithelial cells as possible.
Use IDEIA Chlamydia Specimen Wet Collection Kit for collection of up to 50 specimens. All the components come in self sealed bags for safe transportation.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionIDEIA Chlamydia Specimen Wet Collection Kit
Detectable AnalytesChlamydia
TypeChlamydia Testing Kit
Includes50 x two sterile dacron swabs: one large swab on a plastic shaft (endocervical) and one small swab on a wire and plastic shaft (urethral), 50 x 1mL transport medium
Quantity50 Tests
Unit SizeEach