IDEIA™ PCE Chlamydia Blocking Reagent Kit
Thermo Scientific™
IDEIA™ PCE Chlamydia Blocking Reagent Kit
Verify the positive results obtained with the Thermo Scientific™ IDEIA™ PCE Chlamydia Dual Amplification Assay using this blocking reagent.
Have Questions?
Catalog number S604130-2
Price (INR)
-
Verify the positive results obtained with the Thermo Scientific™ IDEIA™ PCE Chlamydia Kit with Thermo Scientific™ IDEIA™ PCE Chlamydia Blocking Reagent Kit. IDEIA PCE Chlamydia Kit is used to detect genus-specific Chlamydia lipopolysaccharise (LPS) antigen in human urethral and endocervical swabs and male urine samples.
Use IDEIA PCE Chlamydia Blocking Reagents to verify (the specificity of) results of specimens found to be reactive in IDEIA PCE Chlamydia Kit.
IDEIA PCE Chlamydia Blocking Reagents consists of two reagents; a genus-specific murine monoclonal antibody (Blocking Reagent) of different origin to that used in the IDEIA PCE Chlamydia Kit and a murine monoclonal antibody with no antichlamydial activity (Control Reagent).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionIDEIA PCE Chlamydia Blocking Reagent Kit
Detectable AnalytesChlamydia
TypeChlamydia Testing Kit
Includes1mL blocking reagent, 1mL control reagent
Unit SizeEach