IMAGEN™ Chlamydia Trachomatis Specimen Collection Kit
Thermo Scientific™
IMAGEN™ Chlamydia Trachomatis Specimen Collection Kit
Simplify the wet collection and transportation of the urethral and endocervical specimens to be tested with this collection kit.
Have Questions?
Catalog number S610730-2
Price (INR)
-
Simplify the dry collection and transportation of urethral and endocervical specimens to be tested with Thermo Scientific™ IMAGEN™ Chlamydia Kit using Thermo Scientific™ IMAGEN™ Chlamydia Trachomatis Specimen Collection Kit.
Includes: One two-well glass microscope slide, one acetone fixative ampoule, two sterile Dacron-tipped swabs (one standard tip swab for endocervical swabs and one fine swab for urethral swabs).
Principle of the Test
Each IMAGEN™ Chlamydia Collection Pack consists of 20 individual collection kits. Each kit is suitable for the collection of one sample to be tested by the IMAGEN™ Chlamydia Kit for direct detection of Chlamydia spp. in clinical samples.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionIMAGEN Chlamydia Trachomatis Specimen Collection Kit
Detectable AnalytesChlamydia
TypeChlamydia Testing Kit
IncludesTwo-well glass microscope slide, one acetone fixative ampoule, one large Dacron™ sterile swab on plastic shaft, one small Dacron™ sterile swab on stainless steel shaft
Quantity20 collection kits
Unit SizeEach