IMAGEN™ Herpes Simplex Virus Positive Control Slide
Thermo Scientific™
IMAGEN™ Herpes Simplex Virus Positive Control Slide
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide
Have Questions?
Catalog NumberQuantity
S611030-21 Slide
Catalog number S611030-2
Price (INR)
-
Quantity:
1 Slide
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide. A Teflon-coated glass microscope slide with two separate 6mm diameter well areas containing acetone-fixed human fibroblast cells infected with either herpes simplex virus type 1 (HSV 1) or herpes simplex virus type 2 (HSV 2).
Sealed in foil pouch filled with nitrogen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionIMAGEN Herpes Simplex Virus Positive Control Slide
Quantity1 Slide
Unit SizeEach