SYTO™ Green Fluorescent Nucleic Acid Stains
Invitrogen™

SYTO™ Green Fluorescent Nucleic Acid Stains

SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDye TypeQuantity
S7556SYTO 21 Green250 μL
S34854SYTO 9 Green100 μL
S7573SYTO 11 Green250 μL
S7574SYTO 12 Green315 μL
S7575SYTO 13 Green250 μL
S7576SYTO 14 Green250 μL
S7578SYTO 16 Green250 μL
S7559SYTO 24 Green250 μL
S34855SYTO BC Green100 μL
S32703SYTO RNASelect Green100 μL
S7572SYTO Green Dyes 11-14, 16, 21, 24, and 251 Kit (50 μL each)
Catalog number S7556
Price (INR)
24,764.00
Online offer
Ends: 31-Dec-2026
41,272.00
Save 16,508.00 (40%)
Each
Dye Type:
SYTO 21 Green
Quantity:
250 μL
Price (INR)
24,764.00
Online offer
Ends: 31-Dec-2026
41,272.00
Save 16,508.00 (40%)
Each

The SYTO Green Fluorescent Nucleic Acid Stains exhibit bright, green fluorescence upon binding to nucleic acids. These dyes are cell-permeant and can be used to stain RNA and DNA in live cells, dead cells, and bacteria. Because the dye characteristics and staining pattern of the SYTO dyes may vary, we offer 10 different SYTO Green Fluorescent Nucleic Acid Stains available individually along with the SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 (containing SYTO 11–14, 16, 21, 24, and 25) to enable researchers to find the most appropriate green-fluorescent SYTO stain for their system . We also provide the SYTOX Green Nucleic Acid Stain (Cat. No. S7020 and R37109), which is a cell-impermeant green-fluorescent stain used for staining dead cells or performing nuclear counterstaining in fixed cells.

SYTO Green Fluorescent Nucleic Acid Stains are cell-permeant nucleic acid stains that show a large fluorescence enhancement upon binding nucleic acids. The SYTO dyes can be used to stain RNA and DNA in both live and dead eukaryotic cells, as well as in Gram-positive and Gram-negative bacteria.

The SYTO stains share several important characteristics:

  • Permeability to virtually all cell membranes, including mammalian cells and bacteria
  • High molar absorptivity, with extinction coefficients >50,000 cm-1 M-1 at visible absorption maxima
  • Extremely low intrinsic fluorescence, with quantum yields typically <0.01 when not bound to nucleic acids
  • Quantum yields that are typically >0.4 when bound to nucleic acids.

SYTO Green-Fluorescent Nucleic Acid Stains excitation/emission wavelengths

  • SYTO 9—RNA 486/501 nm; DNA 485/498 nm
  • SYTO 11—RNA 510/530 nm; DNA 508/527 nm
  • SYTO 12—RNA 500/519 nm; DNA 499/522 nm
  • SYTO 13—RNA 491/514 nm; DNA 488/509 nm
  • SYTO 14—RNA 521/547 nm; DNA 517/549 nm
  • SYTO 16—RNA 494/525 nm; DNA 488/518 nm
  • SYTO 21—DNA 494/517 nm
  • SYTO 24—DNA 490/515 nm
  • SYTO BC—RNA 487/504 nm; DNA 485/500 nm
  • SYTO RNASelect—RNA 490/530 nm

SYTO dyes differ from each other in one or more characteristics, including cell permeability, fluorescence enhancement upon binding nucleic acids, excitation and emission spectra, DNA/RNA selectivity, and binding affinity. The SYTO dyes are compatible with a variety of fluorescence-based instruments that use laser excitation or a conventional broadband illumination source (e.g., mercury- and xenon-arc lamps). Filter sets that are suitable for imaging cells labeled with fluorescein (FITC), Alexa Fluor 488, or GFP will work well for imaging cells stained with SYTO Green Nucleic Acid Stains.

SYTO nucleic acid stains have been used in diverse applications from staining DNA spotted on microarrays to staining live and fixed cells. The SYTO dyes do not act exclusively as nuclear stains in live cells and should not be equated with DNA-selective compounds such as DAPI (Cat. Nos. D1306, D21490) or Hoechst 33342 (Cat. Nos. H1399, H3570), which stain nuclei in live animal cells. Eukaryotic cells incubated with SYTO dyes generally show cytoplasmic or mitochondrial staining, as well as nuclear staining. The SYTO Green Fluorescent Nucleic Acid Stains have proven valuable in a broad range of research applications as well as used in flow cytometry and fluorescence microscopy.

Specific staining applications for some of the SYTO Green Fluorescent Nucleic Acid Stains

The SYTO 9 stain has been shown to stain live and dead Gram-positive and Gram-negative bacteria. It is a component of the LIVE/DEAD BacLight Bacterial Viability Kits (Cat. Nos. L7007, L7012, L13152).

The SYTO 11 stain (Cat. No. S7573) has been used in conjunction with time-lapse microscopy to examine the cleavage orientation of dividing cells in the developing cerebral cortex.

The SYTO 13 stain (Cat. No. S7575) has been used in combination with propidium iodide (Cat. Nos. P1304, P3566) to monitor glutamate-induced necrosis in cerebellar granule cells.

The SYTO 14 stain (Cat. No. S7576) binds to cytoplasmic RNA, allowing its use in tracking RNA granule transport in living neurons.

Several reports describe the use of SYTO dyes for detecting apoptosis. A series of SYTO nucleic acid stains was screened for the ability to discriminate between apoptotic and non-apoptotic mouse thymocytes, and the SYTO 16 stain (Cat. no. S7578) was found to be optimal for this application. The SYTO 16 stain has also been used with propidium iodide to differentiate live and dead COS-7 cells with a laser-based scanning cytometer.

The SYTO BC Green Fluorescent Nucleic Acid Stain is a mixture of some of our best SYTO dyes for bacterial staining and is a component of the Bacteria Counting Kit (Cat. No. B7277) for bacterial counting in flow cytometry.

The SYTO RNASelect Green Fluorescent Cell Stain is a cell-permeant nucleic acid stain that selectively stains RNA. Although virtually nonfluorescent in the absence of nucleic acids, the SYTO RNASelect stain exhibits bright green fluorescence when bound to RNA (absorption/emission maxima ∼490/530 nm), but only a weak fluorescent signal when bound to DNA. It has been used for staining RNA cargo in exosomes in both live cell samples and isolated exosomes.

SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit

The SYTO Green Fluorescent Nucleic Acid Stain Sampler Kit #1 contains our collection of cell-permeant, green-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 11–14, 16, 21, 24, and 25 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Concentration5 mM solution in DMSO
Detection MethodFluorescence
Dye TypeSYTO 21 Green
EmissionDNA: 517 nm
Excitation Wavelength RangeDNA: 494 nm
For Use With (Application)Fluorescent Labeling, Live Cell Imaging, Microarray
For Use With (Equipment)Fluorescence Microscope, High Content Imager, Flow Cytometer
Product LineSYTO
Quantity250 μL
Shipping ConditionRoom Temperature
Storage BufferDMSO
Volume (Metric)250 μL
Label TypeFluorescent Dye
Product TypeNucleic Acid Stain
Sub Cellular LocalizationNucleic Acids
Unit SizeEach
Contents & Storage
Store in freezer at -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (6)

Citations & References
Abstract
Cellular reactivity to mechanical axonal injury in an organotypic in vitro model of neurotrauma.
Authors:Sieg F, Wahle P, Pape HC
Journal:J Neurotrauma
PubMed ID:10619198
'An in vitro model of traumatic brain injury is described that is based on organotypic cocultures (OTCs) of rat neocortex and thalamus connected by reciprocal axonal projections. Localized mechanical compression of this projection was inflicted with a mechanical device, and the effects on cell viability, axonal morphology, and protein expression ... More
Sarcocystis neurona and Sarcocystis falcatula: monitoring of schizogony in cell culture using fluorescent nuclear labeling.
Authors:Bowman DD, Iordanescu MA, Chou HH, Horton KH
Journal:Vet Parasitol
PubMed ID:11223216
The nuclei of merozoites of Sarcocystis neurona and Sarcocystis falcatula were labeled with the fluorescent marker Syto21. It was shown that the marker would label the parasites and that they would retain the marker throughout schizogony. Thus, there was sufficient marker in the daughter merozoites to make them easily visible ... More
Design of helical proteins for real-time endoprotease assays.
Authors:Steinrücke P, Aldinger U, Hill O, Hillisch A, Basch R, Diekmann S
Journal:Anal Biochem
PubMed ID:11038269
Proteases play a key role in cellular biology and have become priority targets for new pharmaceuticals. Thus, there is a high demand for specific, sensitive, and quick assays to monitor the activity of endoproteases. We designed affinity-tagged helical proteins with unique protease cleavage sites and thus constructed universal, molecularly defined, ... More
Genetically programmed alternative splicing of NEMO mediates an autoinflammatory disease phenotype.
Authors:
Journal:J Clin Invest
PubMed ID:35289316
Single-Cell and Population-Level Analyses Using Real-Time Kinetic Labeling Couples Proliferation and Cell Death Mechanisms.
Authors:
Journal:Dev Cell
PubMed ID:31564612