Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
Fisher Chemicals

Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™

H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Have Questions?
Catalog NumberQuantityPackaging
S9360PB17
also known as S/9360/PB17
2.5 LHDPE Plastic Bottle
Catalog number S9360PB17
also known as S/9360/PB17
HSN 28070010
Price (INR)
9,424.00
Each
Quantity:
2.5 L
Packaging:
HDPE Plastic Bottle
Request bulk or custom format
Price (INR)
9,424.00
Each
Specifications
pH1
ColorColorless
CAS7664-93-9
Melting Point10°C
Boiling Point290°C
View more
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid