Streptavidin, (PE-TR)
Invitrogen™

Streptavidin, (PE-TR)

• Class: RUO• Statement: For research use only. Not for use in diagnostic procedures.• Buffer: PBS• Preservative: 0.1% Sodium AzideViewRead more
Have Questions?
Catalog NumberQuantity
SA10170.5 mL
Catalog number SA1017
Price (INR)
52,127.00
Each
Quantity:
0.5 mL
Price (INR)
52,127.00
Each
Class: RUO
Statement: For research use only. Not for use in diagnostic procedures.
Buffer: PBS
Preservative: 0.1% Sodium Azide

View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferPBS Buffer
Concentration1 mg/mL
Detection MethodFluorescence
Excitation/Emission488/615
Flow Cytometer Laser Lines488 or 561, 488
For Use With (Equipment)Flow Cytometer
Label or DyePE-Texas Red
PackagingTube
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity0.5 mL
Shipping ConditionWet Ice
ConjugatePE-Texas Red
FormSolution
Product LineTexas Red
Unit SizeEach
Contents & Storage
Contains 1 vial of streptavidin, PE-TR conjugate.

Store in refrigerator (2–8°C).