You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Protein Analysis Reagents
▸
Streptavidin-Biotin Binding Products
▸
Streptavidin, (APC-Alexa Fluor™ 750)
Invitrogen™
Streptavidin, (APC-Alexa Fluor™ 750)
Streptavidin, (APC-Alexa Fluor™ 750)
Have Questions?
Catalog Number
Quantity
SA1027
1 mL
Catalog number
SA1027
Price (INR)
48,848.00
Each
Add to cart
Quantity:
1 mL
Price (INR)
48,848.00
Each
Add to cart
Buffer:
PBS
Preservative:
0.1% Sodium Azide
View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer
PBS Buffer
Concentration
1 mg/mL
Detection Method
Fluorescence
Excitation/Emission
600-650/775 nm
Flow Cytometer Laser Lines
633 or 635
For Use With (Equipment)
Flow Cytometer
Label or Dye
APC-Alexa Fluor 750
Packaging
Tube
Product Type
Streptavidin Conjugate (Fluorescent)
Quantity
1 mL
Shipping Condition
Wet Ice
Conjugate
APC-Alexa Fluor 750
Form
Solution
Product Line
Alexa Fluor
Unit Size
Each
Contents & Storage
Contains 1 vial of streptavidin, APC-Alexa Fluor™ 750 conjugate.
Store in refrigerator (2°C to 8°C).