Standard Single-Use Bottle Assembly System, polycarbonate, 3-port cap set, 2 L
Thermo Scientific™
Standard Single-Use Bottle Assembly System, polycarbonate, 3-port cap set, 2 L
This 2-L Thermo Scientific polycarbonate (PC) Single-Use Bottle Assembly System includes a Nalgene Square Polycarbonate Bottle. It is ready toRead more
Have Questions?
Catalog NumberQuantity
SB00005I
also known as SB00005-I
8 Assemblies
Catalog number SB00005I
also known as SB00005-I
Price (INR)
-
Quantity:
8 Assemblies
This 2-L Thermo Scientific polycarbonate (PC) Single-Use Bottle Assembly System includes a Nalgene Square Polycarbonate Bottle. It is ready to use and also includes a 53 B ported closure with one vent filter line and two 1/4-in plugged connection lines.
Specifications
Capacity (English)67.62 oz.
Capacity (Metric)2 L
Closure MaterialPolypropylene
Closure TypeScrew Cap
MaterialPolycarbonate
Quantity8 Assemblies
SterilitySterile
Through the Closure DispensingYes
Workflow StepUpstream sampling. Upstream Validation. Upstream process solution storage.
Product TypeSingle Use Mixer Transfer Assembly
Unit Size8 assemblies
Contents & Storage
Store at room temperature.