CpG Methylated Jurkat Genomic DNA
Thermo Scientific™

CpG Methylated Jurkat Genomic DNA

Thermo Scientific CpG Methylated Jurkat Genomic DNA is human male genomic DNA from Jurkat cells, enzymatically methylated with CpG MethyltransferaseRead more
Have Questions?
Catalog NumberQuantity
SD112115 μg
Catalog number SD1121
Price (INR)
-
Quantity:
15 μg
Thermo Scientific CpG Methylated Jurkat Genomic DNA is human male genomic DNA from Jurkat cells, enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from Jurkat (human acute T-cell leukemia) cells grown to confluency in RPMI media supplemented with 10% bovine fetal serum. Isolated genomic DNA was treated with CpG Methyltransferase, repurified, and dissolved in TE buffer. ≥ 98% CpG methylation level confirmed by bisulfite sequencing.

Highlights

• Pure – DNA free from inhibitors, proteins, and RNA
• Convenient – ready to use 100 ng/μL concentration
• Compatible with downstream applications
• Complete methylation of cytosines in CpG dinucleotides

Applications

As controls in the following:
• Bisulfite sequencing
• Combined Bisulfite Restriction Analysis (COBRA)
• Methylation-sensitive Single-Nucleotide Primer Extension (Ms-SNuPE)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 ng/μL
For Use With (Application)Sequencing
FormLiquid
Includes[other]
Product TypeCpG Methylated Jurkat Genomic DNA
Quantity15 μg
SpeciesHuman
FormatTube
Unit Size15 µg